Learn More
Sino Biological Anti-Cadherin-6/K-cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50985T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Cadherin-6 / CDH6 (Catalog#50985-M08H; P97326; Met1-Ala615). Cadherin-6 / CDH6 specific IgG was purified by Mouse Cadherin-6 / CDH6 affinity chromatography.
Catalog No. 50-241-1468