Learn More
BIOHIPPO Mouse CDHH Microsample
SDP
Catalog No. 5000314907
Encompass
Mouse CDHH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CDHH in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 22 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CDHH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CDHH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CDHH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14907 Supplier BIOHIPPO Supplier No. BHE15211470
Mouse CDHH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CDHH in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 22 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CDHH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CDHH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CDHH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color