Learn More
ACROBiosystems ACROBiosystems Mouse CEACAM-5 Protein, His Tag
Supplier:  ACROBiosystems CE5M52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse CEACAM-5, His Tag (CE5-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Glu 947 (Accession # Q3UKK2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 104.5 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7226