Learn More
Sino Biological Mouse CEACAM1 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 10004H05H1
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CEACAM1 / CD66a (rM CEACAM1 / CD66a; Catalog#50571-M08H; NP_001034274.1; Met1-Gly428) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-237-4380