Learn More
Sino Biological Recombinant Anti-CEACAM-1/CD66a Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 50571R117C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CEACAM1 / CD66a (rM CEACAM1 / CD66a; Catalog#50571-M08H; NP_001034274.1; Met1-Gly428) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-1113