Learn More
Sino Biological Recombinant Mouse CEACAM-5/CD66e Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 54867M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse CEACAM5 (Q3UKK2-1) (Gln35-Glu947) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2962