Learn More
Biomatik Corporation Mouse Cellular Repressor Of E1A Stimulated Genes 1 (CREG1) ELISA Kit
Supplier:  Biomatik Corporation EKU03068
Encompass_Preferred
Mouse Cellular Repressor Of E1A Stimulated Genes 1 (CREG1) ELISA Kit for detecting CREG1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88668. Gene ID: 433375. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9066