Learn More
Biomatik Corporation Mouse Cerberus (CER) CLIA Kit
Supplier:  Biomatik Corporation EKU09832
Encompass_Preferred
Mouse Cerberus (CER) CLIA Kit for detecting CER in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.96pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O55233. Gene ID: 12622. Target Synonyms: CER1; DAND4; Cerberus 1; DAN domain family member 4; Cysteine Knot Superfamily,Homolog (Xenopus Laevis) ; Cerberus-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4644