Learn More
Sino Biological Recombinant Mouse carboxylesterase 2/CES2 Protein (His Tag)
Supplier:  Sino Biological 50515M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CES2 (NP_663578.1) (Met 1-Lys 557) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6780