Learn More
BIOHIPPO Mouse CFB Microsample
SDP
Catalog No. 5000315641
Encompass
Mouse CFB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CFB in serum plasma and other biological fluids It provides a detection range of 6 25-400 g/mL with a sensitivity of 2 49 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CFB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CFB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CFB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15641 Supplier BIOHIPPO Supplier No. BHE15212517
Mouse CFB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CFB in serum plasma and other biological fluids It provides a detection range of 6 25-400 g/mL with a sensitivity of 2 49 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CFB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CFB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CFB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color