Learn More
Sino Biological Mouse CFD MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50539R110100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Adipsin / Complement Factor D (Catalog#50539-M08H; P03953-2; Met1-Ser258).
Catalog No. 50-237-2182