Learn More
BIOHIPPO Mouse CFH Microsample
SDP
Catalog No. 5000315618
Encompass
Mouse CFH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CFH in serum plasma and other biological fluids It provides a detection range of 7 82-500 g/mL with a sensitivity of 4 13 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CFH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CFH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CFH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15618 Supplier BIOHIPPO Supplier No. BHE15212643
Mouse CFH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CFH in serum plasma and other biological fluids It provides a detection range of 7 82-500 g/mL with a sensitivity of 4 13 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CFH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CFH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CFH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color