Learn More
Sino Biological Recombinant Mouse Factor I Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 5A1431M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse CFI (Q61129) (Met1-Val603) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8122