Learn More
Biomatik Corporation Mouse Checkpoint Kinase 1 (CHEK1) ELISA Kit
Supplier:  Biomatik Corporation EKU03088
Encompass_Preferred
Mouse Checkpoint Kinase 1 (CHEK1) ELISA Kit for detecting CHEK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35280. Gene ID: 12649. Target Synonyms: CHK1; Cell cycle checkpoint kinase; Serine/threonine-protein kinase Chk1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6034