Learn More
Biomatik Corporation Mouse Chemerin (CHEM) ELISA Kit
Supplier:  Biomatik Corporation EKU03090
Encompass_Preferred
Mouse Chemerin (CHEM) ELISA Kit for detecting CHEM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9DD06. Gene ID: 71660. Target Synonyms: TIG2; RARRES2; Tazarotene-Induced Gene 2; Retinoic Acid Receptor Responder 2,Tazarotene Induced; RAR-responsive protein TIG2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7518