Learn More
Raybiotech Inc C-Series Mouse Chemokine Antibody Array 1 Kit.
Catalog No. 501948690
Encompass_Preferred
Detects 25 Mouse Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 6Ckine (CCL21), BLC (CXCL13), CTACK (CCL27), CXCL16, Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Fractalkine (CX3CL1), I-TAC (CXCL11), KC (CXCL1), LIX, MCP-1 (CCL2), MCP-5, MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 gamma, MIP-2, MIP-3 beta (CCL19), MIP-3 alpha (CCL20), Platelet Factor 4 (CXCL4), RANTES (CCL5), SDF-1 (CXCL12), TARC (CCL17), I-309 (TCA-3/CCL1), TECK (CCL25)
Catalog No. 50-194-8690 Supplier Raybiotech Inc Supplier No. AAMCHE12
Detects 25 Mouse Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 6Ckine (CCL21), BLC (CXCL13), CTACK (CCL27), CXCL16, Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Fractalkine (CX3CL1), I-TAC (CXCL11), KC (CXCL1), LIX, MCP-1 (CCL2), MCP-5, MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 gamma, MIP-2, MIP-3 beta (CCL19), MIP-3 alpha (CCL20), Platelet Factor 4 (CXCL4), RANTES (CCL5), SDF-1 (CXCL12), TARC (CCL17), I-309 (TCA-3/CCL1), TECK (CCL25)