Learn More
Raybiotech Inc G-Series Mouse Chemokine Array 1 Kit.
Supplier:  Raybiotech Inc GSMCHE11
Encompass_Preferred
Semi-quantitative measurement of 25 mouse chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine (CCL21), BLC (CXCL13), CTACK (CCL27), CXCL16, Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Fractalkine (CX3CL1), I-309 (TCA-3/CCL1), I-TAC (CXCL11), KC (CXCL1), LIX, MCP-1 (CCL2), MCP-5, MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 gamma, MIP-2, MIP-3 alpha (CCL20), MIP-3 beta (CCL19), Platelet Factor 4 (CXCL4), RANTES (CCL5), SDF-1 alpha (CXCL12 alpha), TARC (CCL17), TECK (CCL25)
Catalog No. 50-195-0448