Learn More
Raybiotech Inc Quantibody Mouse Chemokine Array 1 Kit.
Supplier:  Raybiotech Inc QAMCHE12
Encompass_Preferred
Detects 25 Mouse Chemokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: 6Ckine (CCL21), BLC (CXCL13), CTACK (CCL27), CXCL16, Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Fractalkine (CX3CL1), I-309 (TCA-3/CCL1), I-TAC (CXCL11), KC (CXCL1), LIX, MCP-1 (CCL2), MCP-5, MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 gamma, MIP-2, MIP-3 alpha (CCL20), MIP-3 beta (CCL19), Platelet Factor 4 (CXCL4), RANTES (CCL5), SDF-1 alpha (CXCL12 alpha), TARC (CCL17), TECK (CCL25)
Catalog No. 50-195-0870