Learn More
American Research Products Inc Mouse chemokine C C motif li
Supplier:  American Research Products Inc CSBE14298M
Encompass
Mouse chemokine (C-C motif) ligand 1 ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28645