Learn More
Biomatik Corporation Mouse Chemokine (C motif) ligand (XCL1) ELISA Kit
Supplier:  Biomatik Corporation EKU12404
Encompass_Preferred
Mouse Chemokine (C motif) ligand (XCL1) ELISA Kit for detecting C motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47993. Gene ID: 16963. Target Synonyms: Lptn; SCYC1; LTN; ATAC; SCM1-A; SCM1a; LPTN; Lymphotactin; Lymphotaxin; Chemokine C-Motif Ligand 1; Small Inducible Cytokine Subfamily C,Member 1; XC chemokine ligand 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6275