Learn More
American Research Products Inc Mouse chemokine C X C motif
Catalog No. 5000327759
Encompass
Mouse chemokine (C-X-C motif) ligand 17 (CXCL17) ELISA kit from CUSABIO Detection Range 37 5 pg/mL-2400 pg/mL Sensitivity 9 375 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27759 Supplier American Research Products Inc Supplier No. CSBEL006246MO
Mouse chemokine (C-X-C motif) ligand 17 (CXCL17) ELISA kit from CUSABIO Detection Range 37 5 pg/mL-2400 pg/mL Sensitivity 9 375 pg/mL Sample serum plasma tissue homogenates Method Sandwich