Learn More
American Research Products Inc Mouse chemokine C X C motif
Supplier:  American Research Products Inc CSBE17286M
Encompass
Mouse chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating actiVity alpha) (CXCL1) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27743