Learn More
Biomatik Corporation Mouse Chemokine (C-C motif) ligand 6 (CCL6) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11224
Encompass_Preferred
Mouse Chemokine (C-C motif) ligand 6 (CCL6) Mini sample ELISA Kit for detecting C-C motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27784. Gene ID: 20305. Target Synonyms: MRP1; SCYA6; C10; Macrophage Inflammatory Protein Related Protein 1; Small Inducible Cytokine A6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5268