Learn More
Biomatik Corporation Mouse Chemokine C-C-Motif Ligand 1 (CCL1) ELISA Kit
Supplier:  Biomatik Corporation EKU03093
Encompass_Preferred
Mouse Chemokine C-C-Motif Ligand 1 (CCL1) ELISA Kit for detecting CCL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10146. Gene ID: 20290. Target Synonyms: SCYA1; I-309; TCA3; P500; SISe; Small Inducible Cytokine A1; T Lymphocyte Secreted Protein I-309. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5613