Learn More
Biomatik Corporation Mouse Chemokine (C-X-C Motif) Ligand 2 (CXCL2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12179
Encompass_Preferred
Mouse Chemokine (C-X-C Motif) Ligand 2 (CXCL2) Mini sample ELISA Kit for detecting C-X-C Motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10889. Gene ID: 20310. Target Synonyms: GROb; SCYB2; MIP2; GRO2; MIP2a; MGSAb; CINC2a; HSF; Macrophage inflammatory protein 2-alpha; Hematopoietic synergistic factor; Growth Regulated Oncogene Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6593