Learn More
Biomatik Corporation Mouse Chemokine C-X-C-Motif Ligand 16 (CXCL16) ELISA Kit
Supplier:  Biomatik Corporation EKU09023
Encompass_Preferred
Mouse Chemokine C-X-C-Motif Ligand 16 (CXCL16) ELISA Kit for detecting CXCL16 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BSU2. Gene ID: 66102. Target Synonyms: SR-PSOX; CXCLG16; SRPSOX; Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein; Small-inducible cytokine B16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8575