Learn More
BIOHIPPO Mouse CHGA Microsample
SDP
Catalog No. 5000314993
Encompass
Mouse CHGA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGA in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CHGA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CHGA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CHGA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14993 Supplier BIOHIPPO Supplier No. BHE15211338
Mouse CHGA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGA in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CHGA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CHGA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CHGA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color