Learn More
BIOHIPPO Mouse CHGB Microsample
SDP
Catalog No. 5000313986
Encompass
Mouse CHGB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGB in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CHGB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CHGB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CHGB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13986 Supplier BIOHIPPO Supplier No. BHE15211123
Mouse CHGB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGB in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CHGB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CHGB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CHGB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color