Learn More
Sino Biological Recombinant Mouse CHL-1/L1CAM-2 Protein (His Tag) 500
Supplier:  Sino Biological 51176M08H500
Encompass_Preferred
A DNA sequence encoding the mouse CHL1(XP_017176866.1) (Met1-Gln1027) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6842