Learn More
Biomatik Corporation Mouse Cholecystokinin 8 (CCK8) ELISA Kit
Supplier:  Biomatik Corporation EKU03140
Encompass_Preferred
Mouse Cholecystokinin 8 (CCK8) ELISA Kit for detecting CCK8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.01pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P09240. Gene ID: 12424. Target Synonyms: Sincalide; Cholecystokinin 8, Octapeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5146