Learn More
Biomatik Corporation Mouse Cholecystokinin A Receptor (CCKAR) ELISA Kit
Supplier:  Biomatik Corporation EKU03147
Encompass_Preferred
Mouse Cholecystokinin A Receptor (CCKAR) ELISA Kit for detecting CCKAR in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08786. Gene ID: 12425. Target Synonyms: CCK-A; CCK1; CCKRA; CCK1R; Cholecystokinin Type 1 Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7795