Learn More
Raybiotech Inc Mouse Chordin ELISA Kit
Supplier:  Raybiotech Inc ELMCHORDIN1
Encompass_Preferred
For Cell Culture Supernatants, EDTA and/or Citrate treated plasma, and Serum samples. Heparin is not recommended. Sensitivity: 49 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse Chordin.
Catalog No. 50-194-9949