Learn More
Biomatik Corporation Mouse Chordin Like Protein 2 (CHRDL2) ELISA Kit
Supplier:  Biomatik Corporation EKU03172
Encompass_Preferred
Mouse Chordin Like Protein 2 (CHRDL2) ELISA Kit for detecting CHRDL2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VEA6. Gene ID: 69121. Target Synonyms: BNF1; CHL2; Breast Tumor Novel Factor 1; Chordin-related protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7186