Learn More
Biomatik Corporation Mouse Chromatin Assembly Factor 1, Subunit B (CHAF1B) ELISA Kit
Supplier:  Biomatik Corporation EKU03176
Encompass_Preferred
Mouse Chromatin Assembly Factor 1, Subunit B (CHAF1B) ELISA Kit for detecting CHAF1B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.052ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D0N7. Gene ID: 110749. Target Synonyms: CAF-1; CAF-IP60; CAF1; CAF1A; CAF1P60; MPHOSPH7; MPP7; M-Phase Phosphoprotein 7; Chromatin Assembly Factor I,p60 subunit; M-phase phosphoprotein 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6879