Learn More
Biomatik Corporation Mouse Chromogranin A (CHGA) ELISA Kit
Supplier:  Biomatik Corporation EKU09052
Encompass_Preferred
Mouse Chromogranin A (CHGA) ELISA Kit for detecting CHGA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 48pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26339. Gene ID: 12652. Target Synonyms: CGA; SP-I; Parathyroid Secretory Protein 1; Vasostatin; Pancreastatin; Parastatin; Pituitary secretory protein I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7255