Learn More
Biomatik Corporation Mouse Chromogranin B (CHGB) ELISA Kit
Supplier:  Biomatik Corporation EKU03182
Encompass_Preferred
Mouse Chromogranin B (CHGB) ELISA Kit for detecting CHGB in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16014. Gene ID: 12653. Target Synonyms: SCG1; CgB; SgI; Secretoneurin 1; Secretogranin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6410