Learn More
Biomatik Corporation Mouse Chromosome 4 Open Reading Frame 48 (C4orf48) ELISA Kit
Supplier:  Biomatik Corporation EKU12104
Encompass_Preferred
Mouse Chromosome 4 Open Reading Frame 48 (C4orf48) ELISA Kit for detecting C4orf48 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3UR78. Gene ID: 381633. Target Synonyms: Neuropeptide-like protein C4orf48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7226