Learn More
Biomatik Corporation Mouse Ciliary Neurotrophic Factor (CNTF) CLIA Kit
Supplier:  Biomatik Corporation EKU09601
Encompass_Preferred
Mouse Ciliary Neurotrophic Factor (CNTF) CLIA Kit for detecting CNTF in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.44pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P51642. Gene ID: 12803. Target Synonyms: HCNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7214