Learn More
American Research Products Inc Mouse Ciliary Neurotrophic Fac
Catalog No. 5000328839
Encompass
Mouse Ciliary Neurotrophic Factor CNTF ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-28839 Supplier American Research Products Inc Supplier No. CSBE07312M
Mouse Ciliary Neurotrophic Factor CNTF ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich