Learn More
American Research Products Inc Mouse circulating immune compl
Catalog No. 5000328612
Encompass
Mouse circulating immune compleX (CIC) ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28612 Supplier American Research Products Inc Supplier No. CSBE16553M
Mouse circulating immune compleX (CIC) ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich