Mouse Claudin-18.2 full length protein presented on virus-like particles (VLPs) for immunization and antibody screening. VLPs display conformationally intact membrane protein in a native-like lipid environment, with compact particle diameter suitable for antigen presentation. Supplied as a filtered solution in PBS with arginine and trehalose.
- Format: virus-like particles presenting full-length mouse claudin-18.2.
- Amount: 100 μg per vial.
- Expression system: produced in human 293 cells (HEK293).
- Amino acid range: Met 1-Val 264.
- Supplied buffer: PBS with arginine and trehalose; 0.2 μm filtered.
- Storage: store at -70°C or lower; stable for 12 months; avoid repeated freeze-thaw.
- Applications: suitable for immunization and antibody screening.
- Particle size: DLS mean peak radius ~65-85 nm; VLP diameter ~100-300 nm.
- Purity indicator: DLS shows >95% intensity.