Learn More
Biomatik Corporation Mouse Claudin 3 (CLDN3) ELISA Kit
Supplier:  Biomatik Corporation EKU03208
Encompass_Preferred
Mouse Claudin 3 (CLDN3) ELISA Kit for detecting CLDN3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z0G9. Gene ID: 12739. Target Synonyms: C7orf1; CPE-R2; CPETR2; HRVP1; RVP1; Clostridium Perfringens Enterotoxin Receptor 2; Ventral Prostate.1-Like Protein; CPE-Receptor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5308