Learn More
BIOHIPPO Mouse CLEC10A Microsample
SDP
Catalog No. 5000314378
Encompass
Mouse CLEC10A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLEC10A in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CLEC10A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CLEC10A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CLEC10A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14378 Supplier BIOHIPPO Supplier No. BHE15210775
Mouse CLEC10A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLEC10A in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CLEC10A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CLEC10A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CLEC10A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color