Learn More
Sino Biological Anti-CLEC10A/CD301 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50707T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CLEC10A / MGL1 / CD301 (Catalog#50707-M07H; NP_001191181.1; Gln58-Ser305). CLEC10A / MGL1 / CD301 specific IgG was purified by Mouse CLEC10A / MGL1 / CD301 affinity chromatography.
Catalog No. 50-255-3135