Learn More
Sino Biological Anti-CLEC10A/CD301 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50707RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CLEC10A / MGL1 / CD301 (Catalog#50707-M07H; NP_001191181.1; Gln58-Ser305). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3136