Learn More
ACROBiosystems Mouse CLEC12A / MICL / CLL-1 Protein, His Tag
Catalog No. 502054843
Encompass_Preferred
Recombinant Protein;1MG;Mouse CLEC12A, His Tag (CLA-M5244) is expressed from human 293 cells (HEK293). It contains AA Tyr 65 - Arg 267 (Accession # Q504P2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.7 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4843 Supplier ACROBiosystems Supplier No. CLAM52441MG
Recombinant Protein;1MG;Mouse CLEC12A, His Tag (CLA-M5244) is expressed from human 293 cells (HEK293). It contains AA Tyr 65 - Arg 267 (Accession # Q504P2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.7 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.