Learn More
Sino Biological Recombinant Mouse CLL-1/CLEC12A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50558M07H100
Encompass_Preferred
A DNA sequence encoding the Mouse CLEC12A (NP_808354.1) (Tyr65-Arg267) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-241-2139