Learn More
Sino Biological Recombinant Mouse DICR/CLEC4A Protein (ECD, His Tag)
Supplier:  Sino Biological 50945M07H100
Encompass_Preferred
A DNA sequence encoding the mouse CLEC4A (NP_001163803.1) (Gln70-Leu238) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3046