Learn More
Sino Biological Anti-Dectin-2/CLEC6A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50267T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Dectin-2/CLEC6A (rM Dectin-2/CLEC6A; Catalog#50267-M08H; NP_064385.1; Ile44-Leu209). Dectin-2/CLEC6A specific IgG was purified by Mouse Dectin-2/CLEC6A affinity chromatography.
Catalog No. 50-255-3320