Learn More
Sino Biological Recombinant Anti-Dectin-1/CLEC7A Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50233R010100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Dectin-1 / CLEC7A (rM Dectin-1 / CLEC7A; Catalog#50233-M07H; NP_064392.2; Phe69-Leu244).
Catalog No. 50-255-3355